Benchmark Scientific™ MyFuge 12 Benchtop Centrifuge for Microtubes and PCR tubes
Supplier: Alkali Scientific
RS7183
104029-042EA 461.52 USD
104029-042
Benchmark Scientific™ MyFuge 12 Benchtop Centrifuge for Microtubes and PCR tubes
Centrifuges Microcentrifuges
The MyFuge12 mini centrifuge features a combined rotor that spins up to 12 microtubes and 4 strip tubes all at once.
  • COMBI-Rotor for tubes and strips
  • Twice the capacity of most mini centrifuges
  • Near silent operation
  • Starts and stops with closing/opening of the lid
  • Conserves valuable bench space
Unlike traditional mini centrifuge, the new MyFuge™ 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor™ is all that is required for running 12 microtubes and/or 4 PCR strips.With a fixedMore Product Information