Sodium dihydrogen phosphate 1M, CellPro™

Supplier: Alkali Scientific

DS0328
104029-028EA 49.22 USD
104029-028
Sodium dihydrogen phosphate 1M, CellPro™
Sodium dihydrogen phosphate

CellPro™ Sodium Phosphate Solution offers researchers a high-quality reagent for molecular biology applications. Crafted to stringent standards of purity and sterility, this solution ensures reliable performance in experimental procedures. With its sterile filtered formulation, CellPro™ Sodium Phosphate Solution provides researchers with confidence and peace of mind in their work, facilitating successful outcomes in molecular biology studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: NaH₂PO₄
MDL Number: MFCD00003527
CAS Number: 7558-80-7
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR