Streptomycin solution 10 mg/ml, CellPro™

Supplier: Alkali Scientific

DS0178
104028-992EA 83.06 USD
104028-992
Streptomycin solution 10 mg/ml, CellPro™
Streptomycin sulphate

CellPro™ Streptomycin Solution provides researchers with a superior antibiotic option for molecular biology applications. Crafted to meticulous standards of purity and sterility, this solution ensures reliable inhibition of bacterial growth in experimental procedures. With its sterile filtered formulation and concentration of 10mg/mL in a 10mL volume, CellPro™ Streptomycin Solution offers researchers confidence and peace of mind in their molecular biology endeavors.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
MDL Number: MFCD00037023
CAS Number: 3810-74-0
UN: 3248

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR