Ampicillin solution 10 mg/ml, CellPro™

Supplier: Alkali Scientific

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

DS0198
104028-972EA 124.6 USD
104028-972
Ampicillin solution 10 mg/ml, CellPro™
Ampicillin sodium salt

Alkali Scientific™ Ampicillin Solution offers researchers a high-quality antibiotic solution for molecular biology applications. Crafted to stringent standards of purity and sterility, this sterile filtered solution ensures reliable and consistent inhibition of bacterial growth in experimental procedures. Ideal for selective bacterial culture and plasmid maintenance, Alkali Scientific™ Ampicillin Solution guarantees optimal results and peace of mind in your laboratory work.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
MDL Number: MFCD00064313
CAS Number: 69-52-3
UN: 3248

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR