Glycerol, CellPro™

Supplier: Alkali Scientific

DB0268
104028-854EA 49.22 USD
104028-854
Glycerol, CellPro™
Glycerine

Introducing CellPro™ Glycerol, an essential component meticulously crafted by Alkali Scientific™ to meet the exacting standards of molecular biology research. With a generous volume of 500mL, this high-quality glycerol solution offers unparalleled purity and reliability for a wide range of laboratory applications. Whether used as a cryoprotectant, a component of buffers, or for protein stabilization, CellPro™ Glycerol ensures optimal performance and reproducibility in your experiments. Trust CellPro™ Glycerol to deliver consistent results, enabling you to advance your research with confidence.


  • For sample preparation and gel formation for polyacrylamide gel electrophoresis
  • Used in casting gradient gels, as a protein stabilizer and storage buffer component

Formula: HOCH₂CH(OH)CH₂OH
MDL Number: MFCD00004722
CAS Number: 56-81-5
UN: 1219

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR