Neomycin sulfate solution 8-12 mg/ml, CellPro™, sterile

Supplier: Alkali Scientific

Synonyms: Neomycin, Fradiomycin sulfate, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate, Fradiomycin sulphate, Neomycin sulphate salt, Neomycin sulfate salt, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulfate

DS0183
104028-748EA 33.83 USD
104028-748
Neomycin sulfate solution 8-12 mg/ml, CellPro™, sterile
Neomycin sulphate

CellPro™ Neomycin Solution offers researchers a high-quality antibiotic solution for molecular biology applications. Crafted to stringent standards of purity and sterility, this solution ensures reliable and consistent inhibition of bacterial growth in experimental procedures. With its sterile filtered formulation, CellPro™ Neomycin Solution provides researchers with confidence and peace of mind in their work, facilitating successful outcomes in molecular biology studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
MW: 908.87 g/mol
MDL Number: MFCD00150827
CAS Number: 1405-10-3
UN: 3248

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR