MyFuge™ Mini Microcentrifuge
Supplier: Alkali Scientific
C9310
104028-684EA 332.29 USD
104028-684
MyFuge™ Mini Microcentrifuge
Centrifuges Microcentrifuges
Efficiently spin down microcentrifuge tubes with the MyFuge™ Mini Microcentrifuge.
  • Eight position microtube rotor
  • Four position PCR strip rotor
  • Storage compartment for PCR rotor
  • Near silent operation
  • Starts and stops in seconds
  • Saves valuable bench space
The MyFuge™ Mini microcentrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. An increased capacity, eight-position microtube rotor is included and comes installed. Also included is a second rotor for 0.2 ml PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use, storage in this compartment prevents loss or misplacement of the rotor.
Ordering information: With 2 years warranty.