CellPro™ Reducing Adapter
Supplier: Alkali Scientific
MT4200
104028-486EA 63.06 USD
104028-486
CellPro™ Reducing Adapter
Adapters Adapters, Conical Ground Joints
Seamlessly adapt laboratory processes with the CellPro™ Reducing adapter.
  • For use with 15 ml, 17×100 mm, 12×75 mm, and 5 ml conical tubes
The CellPro™ Reducing adapter is an essential accessory designed to enhance the versatility of a laboratory centrifuge. This adapter providing flexibility for different experimental needs. With a pack containing 25 adapters, users can streamline centrifugation processes and ensure efficient sample processing in research workflows.
Designed with precision and durability in mind, the CellPro™ Reducing adapter is an indispensable tool for any laboratory seeking reliable and consistent centrifugation performance.