Radiant™ Hot Start Taq Polymerase
Supplier: Alkali Scientific
C325
104028-464EA 810.75 USD
104028-464
Radiant™ Hot Start Taq Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Unlock precise amplification with Radiant™ Hot start Taq polymerase.
  • New-generation 10X PCR buffer, MgCl₂ and enhancers for maximum PCR efficiency and reaction speed
  • Robust PCR performance across a wide range of DNA templates including multiplex assays and problematic templates
  • High-yields with amplicons up to 5 Kb with standard or fast cycling
Radiant™ Hot-start Taq polymerase Radiant™ Hot-start Taq DNA polymerase is a highly purified, high performance Hot-start DNA polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA and crude samples. The Hot-start technology is based on a new-generation PCR buffer formulation in conjunction with a antibody-mediated chemistry. Radiant™ Hot-start Taq polymerase exhibits 5´ to 3´ DNA polymerase activity with an error-rate of wild-type Taq (2.0×10-5). The polymerase is ideal for genotyping, colony PCR, multiplexing, low-copy assays, and the amplification of challenging targets susceptible to mispriming.
Radiant™ Hot start Taq DNA polymerase is engineered for robust, superior PCR and is supplied with a highly optimized, new-generation 5X buffer system which provides exceptional sensitivity and ease of use. Storage Radiant™ Hot-Start Taq Polymerase is shipped on blue or dry ice and should be stored at –20 °C upon receipt. Excessive freeze/thawing should be avoided. When stored as specified, Radiant™ Hot-Start Taq DNA Polymerase is stable for 12 months from date of receipt. The Kit may also be stored at 4 °C for 1 month. Important Considerations Radiant™ 10X HS Taq reaction buffer Template: For complex genomic DNA, we suggest the use of 5 to 500 ng per reaction; For cDNA or plasmid DNA, please use <100 ng per reaction. Primers: Primers should have a predicted melting temperature of around 60 °C, using default Primer 3 settings.
The final primer concentration in the reaction should be between 0.2 and 0.6 μM. Annealing: We recommend performing a temperature gradient to determine the optimal annealing temperature. Alternatively, we suggest a 55 °C annealing temperature. Increase in 2 °C increments if non-specific products are present. Extension: Optimal extension is achieved at 72 °C. The optimal extension time is dependent on amplicon length and complexity. 15 seconds per kilobase (Kb) is recommended for amplification from eukaryotic genomic DNA or cDNA. For shorter amplicons, a 1 second extension is sufficient.
For multiplex PCR, we suggest an initial annealing temperature gradient from 55 to 65 °C in order to determine the highest level of specificity. In addition, we recommend an initial extension time of 90 seconds and greater to maximize yield and specificity. Quality control Radiant™ Hot-start Taq polymerase is tested extensively for robust activity, processivity, efficiency, heat activation, sensitivity, absence of nuclease contamination and absence of nucleic acid contamination. Radiant™ Hot-start Taq polymerase is manufactured under a comprehensive quality management system, following ISO 9001:2008 standards.
Caution: Limitations of use: This product is intended for research purposes only and is not intended for any animal or human therapeutic use.