ProCycle™ 0.1 ml PCR 8-Strip Tubes, Clear
Supplier: Alkali Scientific
PCN7055
104028-414EA 693.83 USD
104028-414
ProCycle™ 0.1 ml PCR 8-Strip Tubes, Clear
Tubes PCR Tubes
Conduct precise PCR assays with Alkali Scientific™ ProCycle™ 0.1 ml PCR 8-Strip Tubes, featuring clear material and available in packs of 125.
  • Fast® Cycler and Lightcycler® compatible
  • Angularly attached caps on UltraFlux® i and SnapStrip® tubes prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack
  • Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing
  • Clear tubes provide high clarity for visualizing contents
  • White tubes offer enhanced fluorescence reflection and the optically-clear caps, thanks to proprietary two-material manufacturing, ensure that the fluorescence signal is passed through uninhibited, creating the perfect environment for consistent and reliable qPCR performance
  • For use in pharmaceutical industry, food industry and molecular biology
  • Manufactured from high-quality, virgin polypropylene to ensure purity and provide e×cellent optical clarity
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free
Maximize PCR efficiency with ProCycle™ 0.1 ml PCR 8-Strip Tubes. Designed for high-throughput applications, these clear tubes offer precise sample volume control and efficient heat transfer for optimal PCR performance. With a capacity of 0.1 ml per tube, they are ideal for small-scale PCR reactions and sample conservation. Each pack contains 125 tubes, providing ample supply for your PCR experiments.