Magnesium chloride solution 1M, CellPro™

Supplier: Alkali Scientific

DS0213 DS0189
104027-244EA 110.75 USD
104027-244 104028-356
Magnesium chloride solution 1M, CellPro™
Magnesium chloride

CellPro™ Magnesium Chloride 1M Solution provides researchers with a high-quality source of magnesium ions for molecular biology applications. Crafted to stringent standards of purity and sterility, this solution ensures reliable performance in experimental procedures. With its sterile filtered formulation, CellPro™ Magnesium Chloride Solution offers researchers confidence and peace of mind in their work, facilitating successful outcomes in molecular biology studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: MgCl₂
MDL Number: MFCD00011106
CAS Number: 7786-30-3
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR