Newborn Calf Serum, 500 ml Storage: 20 °C Virus Screened Mycoplasma Tested 0.2 µm Filtered

Supplier: Alkali Scientific
FB56
104028-182EA 141.52 USD
104028-182
Newborn Calf Serum, 500 ml Storage: 20 °C Virus Screened Mycoplasma Tested 0.2 µm Filtered
Sera (Serum)
Promote cell proliferation with Alkali Scientific™ Newborn calf serum, available in a 500 ml volume for optimal cell culture conditions.

  • Triple 0.1 µm sterile-filtered
  • Mycoplasma-tested and virus-screened
  • Ships on dry ice; Store frozen at −20 to −10 °C

CellPro™ Newborn Calf Serum is a premium-quality serum product specifically designed to support cell culture applications in biomedical research. Sourced from the blood of healthy newborn calves and processed under stringent quality control measures, this serum product offers researchers a reliable and consistent source of growth factors, hormones, and nutrients necessary for cell growth and proliferation. With its carefully optimized composition and sterile packaging, CellPro™ Newborn Calf Serum provides researchers with confidence in the quality and performance of their cell culture experiments, ensuring robust and reproducible results. Whether used for routine cell culture maintenance, cell-based assays, or specialized research applications, CellPro™ Newborn Calf Serum stands as a trusted choice for researchers seeking high-quality and reliable serum products for their laboratory work.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR