In situ Adapter for Use with Combination Blocks for Thermal Cycler
Supplier: Alkali Scientific
T5000-A-IS
104028-062EA 727.68 USD
104028-062
In situ Adapter for Use with Combination Blocks for Thermal Cycler
Thermal Cyclers
Enhance thermal cycling experiments with the in situ adapter, designed for use with combination blocks in your thermal cycler.
  • In situ adapter fits on the TC 9639 thermal cycler block and holds four standard slides
This specialized adapter, compatible with combination blocks for thermal cyclers, facilitates precise and efficient in situ procedures. Designed for seamless integration with your existing equipment, it ensures reliable and reproducible results in molecular biology applications.