Penicillin : Streptomycin solution, 10,000 I.U. Penicillin, 10,000 ug/ml Streptomycin

Supplier: Alkali Scientific

FB84
104028-038EA 49.22 USD
104028-038
Penicillin : Streptomycin solution, 10,000 I.U. Penicillin, 10,000 ug/ml Streptomycin
Penicillin/Streptomycin

CellPro™ Penicillin Streptomycin (Pen-Strep) Solution offers researchers a high-quality antibiotic solution for cell culture applications in biomedical research. With a concentration of 10,000 µg/mL, this solution provides potent and effective antimicrobial activity against a wide range of gram-positive and gram-negative bacteria, ensuring reliable inhibition of bacterial contamination in cell culture experiments. Formulated with precision and purity in mind, CellPro™ Penicillin Streptomycin Solution maintains sterility and stability throughout its shelf life, offering researchers a dependable and consistent antibiotic option for their cell culture work. Whether used for routine cell culture maintenance or specialized cell-based assays, CellPro™ Penicillin Streptomycin Solution empowers researchers with confidence in the integrity and reliability of their experimental results.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR