Gentamicin solution 50 mg/ml, CellPro™

Supplier: Alkali Scientific

DS0181
104027-924EA 101.52 USD
104027-924
Gentamicin solution 50 mg/ml, CellPro™

CellPro™ Gentamycin Solution stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution offers researchers a reliable antibiotic option for cell culture applications. With its sterile filtered formulation, CellPro™ Gentamycin Solution ensures consistent and reproducible results in experimental procedures, providing researchers with peace of mind and confidence in their work.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: C₂₁H₄₃N₅O₇
CAS Number: 1403-66-3
UN: 3248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR