Autoclaving Jar
Supplier: Alkali Scientific
AJ2080
104027-788EA 19.98 USD
104027-788
Autoclaving Jar
Jars General Use Jars
Optimize autoclaving processes with this durable 120×80 mm autoclaving jar.
  • Transparent polypropylene
The autoclaving jar is a durable and heat-resistant container designed for sterilization processes in autoclaves. With dimensions of 120×80 mm, it offers ample space for holding small laboratory instruments, tools, or materials during autoclaving. Constructed from sturdy materials capable of withstanding high temperatures and pressure, this jar ensures effective sterilization of contents, promoting safety and hygiene in the laboratory.