ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Dome Strip Caps
Supplier: Alkali Scientific
PC7063
104027-704EA 110.75 USD
104027-704
ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Dome Strip Caps
Tubes PCR Tubes
Efficiently conduct PCR experiments with Alkali Scientific™ ProCycle™ 0.2 ml 8-Strip PCR tubes featuring separate dome strip caps, available in packs of 1000.
  • 0.2 ml strip tubes with separate domed caps
  • Ultra-thin walls
  • Positive sealing
  • Fits standard spacing for 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Experience versatility and reliability at its finest with the ProCycle™ 0.2 ml 8-Strip PCR tubes and separate dome strip caps. Engineered for rapid thermal transfer and optimal sample integrity, these tubes and caps guarantee consistent performance, empowering researchers to achieve reproducible results with ease, even in high-throughput PCR assays. With a generous pack size of 1000 tubes, researchers can strea mline their experiments without compromising on reliability or efficiency.