ProCycle™ SnapStrip 8-Strip Tubes with Single EZCut™ Connectors and Individually Attached Flat Caps
Supplier: Alkali Scientific
PC7061-ONE
104027-684EA 110.75 USD
104027-684
ProCycle™ SnapStrip 8-Strip Tubes with Single EZCut™ Connectors and Individually Attached Flat Caps
Tubes PCR Tubes
Streamline PCR workflow with Alkali Scientific™ ProCycle™ SnapStrip 8-Strip tubes featuring single EZCut™ connectors and individually attached flat caps, providing clear visibility of samples.
  • 0.2 ml 8-Strip PCR tubes with attached caps (120 strip × 8, 960/pk)
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Optical flat caps
  • Certified RNAse, DNAse and pyrogen free
Experience enhanced convenience and efficiency in your PCR workflow with Alkali Scientific™ ProCycle™ SnapStrip 8-Strip tubes. These tubes feature single EZCut™ connectors that allow for easy separation into individual tubes, eliminating the need for additional tools and minimizing the risk of sample cross-contamination. Each tube is equipped with an individually attached flat cap, providing a secure seal to prevent sample evaporation and contamination during thermal cycling. The clear design of the tubes enables easy visualization of your samples, ensuring accurate pipetting and tracking of reactions.