96-Well Deep Well Plate, 0.5 ml, V Conical Bottom, Square Well, Sterile
Supplier: Alkali Scientific
TL456
104027-568EA 226.14 USD
104027-568
96-Well Deep Well Plate, 0.5 ml, V Conical Bottom, Square Well, Sterile
Microplates Deep Well Microplates
Conduct efficient sample processing with CellPro™ 0.5 ml 96-well deep well plate, featuring a V conical bottom and square well design, suitable for the KingFisher extraction platform, and available sterile in a pack of 100.
  • Made of medical grade polypropylene
  • Compatible with Thermo Fisher KingFisher™ instruments
  • Low binding assures maximum recovery
This 0.5 ml 96-well deep well plate, designed for use with the KingFisher extraction platform. Featuring a V conical bottom and square wells, these plates ensure efficient sample recovery and processing. Manufactured under sterile conditions, they are ready for immediate use in your laboratory workflows. With a capacity of 0.5 ml per well, these plates provide ample space for sample handling, making them ideal for high-throughput applications such as nucleic acid extraction, PCR setup, and sample storage.