0.5 ml 96-Well, Deep Well Plates, V Conical Bottom, Square Well, Sterile
Supplier: Alkali Scientific
MPD22-S
104027-410EA 396.91 USD
104027-410
0.5 ml 96-Well, Deep Well Plates, V Conical Bottom, Square Well, Sterile
Microplates Deep Well Microplates
Ensure sterile sample handling with 0.5 ml 96-Well Deep Well Plates, featuring a V Conical Bottom and square well design, available in a sterile case of 50.
  • Virgin, heat-resistant, medical-grade polypropylene plates
  • RNase, DNase and pyrogen free
  • Minimal binding and liquid retention
  • Black printed alpha-numeric reference grids
  • Autoclavable; sterile plates are sterilized by gamma irradiation in accordance with ISO 13485
  • Temperature range: –86 to +82.2 °C
These 96-Well Deep Well Plates are engineered for efficient sample processing and storage in high-throughput applications. Featuring a V conical bottom and square well design, this deep well plate maximizes sample recovery and mixing, making it ideal for a wide range of laboratory procedures, including cell culture, compound screening, and sample storage.
Constructed from high-quality polypropylene material, it offers excellent chemical resistance and compatibility with a variety of laboratory reagents and solvents. Each plate is sterile and RNase/DNase-free, ensuring the integrity of samples and reliable experimental results. With alphanumeric labeling and raised rims for easy sample identification and handling, the 96-Well Deep Well Plate provides researchers with a dependable platform for high-throughput sample processing in molecular biology, genomics, and proteomics research.