Radiant™ SYBR Green Fluorescein qPCR Kit
Supplier: Alkali Scientific
QS3001
104027-400EA 78.45 USD
104027-400
Radiant™ SYBR Green Fluorescein qPCR Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Detect target DNA efficiently with Radiant™ SYBR green fluorescein qPCR Kit, offering reliable performance for quantitative PCR assays.
  • Next-generation PCR buffer formulations for maximum PCR efficiency and reaction speed
  • Novel hot-start chemistry for improved specificity and sensitivity
  • Broad range detection for increased reliability in low-copy assays
  • Versatile 2X mixes, suitable for both ultra-fast and standard cycling methods
  • Compatible with BioRad® iCycler™, MyIQ™ and IQ5™
Radiant™ Green Fluorescein qPCR Kits, a high-performance 2X ready-to-use real-time PCR reagents designed for quantitative PCR. These kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The buffer system allows highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low-copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Green Fluorescein qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection in combination with fluorescein for optional use with certain qPCR instruments.
Instrument Compatibility: The Radiant™ Green Fluorescein qPCR Kit has been optimized for use with intercalating-dye based real-time PCR. Each of these instruments has the capacity to analyze qPCR data with the passive reference signal either on or off.