12-Strip Tubes and 12-Strip Flat Caps Packed Together
Supplier: Alkali Scientific
PC7014
104027-398EA 138.45 USD
104027-398
12-Strip Tubes and 12-Strip Flat Caps Packed Together
Tubes PCR Tubes
Simplify laboratory workflow with the 12-strip tubes and flat caps combo pack. This convenient pack includes 80 strips, each containing 12 tubes and 12 flat caps providing you with a total of 960 tubes and caps.
  • Angularly attached caps prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack
  • Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing
  • Clear tubes provide high clarity for visualizing contents
  • For use in pharmaceutical industry, food
More Product Information