Magnesium sulfate solution 1M, CellPro™

Supplier: Alkali Scientific

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

DS0191
104027-176EA 29.22 USD
104027-176
Magnesium sulfate solution 1M, CellPro™
Magnesium sulphate

CellPro™ Magnesium Sulfate Solution offers researchers a reliable source of magnesium ions for molecular biology applications. Crafted to meticulous standards of purity and sterility, this solution ensures consistent performance in experimental procedures. With its sterile filtered formulation, CellPro™ Magnesium Sulfate Solution provides researchers with confidence and peace of mind in their work, facilitating successful outcomes in molecular biology studies.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127
MDL Number: MFCD00011110
CAS Number: 7487-88-9
UN: 3248
Merck Index: 12,05731

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR