ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Domed Cap
Supplier: Alkali Scientific
PC7066
104027-048EA 110.75 USD
104027-048
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Domed Cap
Tubes PCR Tubes
Enhance sample visibility with Alkali Scientific™ ProCycle™ 0.2 ml clear 8-Strip tubes featuring attached hinged domed caps, available in packs of 1000.
  • 0.2 ml 8-Strip PCR tubes with attached dome caps
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Streamline laboratory operations with the ProCycle™ 0.2 ml clear 8-Strip PCR tubes and attached hinged domed caps. Designed for rapid thermal transfer and secure sealing, each pack of 1000 tubes ensures uninterrupted experimentation, allowing researchers to accelerate their scientific progress with ease and efficiency. With their transparent design, researchers can easily monitor samples while enjoying the convenience of attached hinged domed caps.