OmniPette™ Single Channel Pipette
Supplier: Alkali Scientific
P9280-50U
104026-792EA 209.22 USD
104026-792
OmniPette™ Single Channel Pipette
Pipettes Micropipettes
Achieve accurate dispensing with OmniPette single channel pipette, offering a volume range of 5 to 50 μl.
  • Stainless steel pistons and teflon seals
  • Superior accuracy and reproducibility
  • Adjustable volume and locking wheel
  • Full volume range (0.1 µl to 10 ml)
  • Outstanding user comfort
  • Fully autoclavable
  • 3 year warranty
Experience unparalleled precision and versatility with the OmniPette™ Single channel pipette. This pipette offers exceptional performance for a wide range of liquid handling tasks in the laboratory. Its ergonomic design ensures comfortable operation during extended use, while the adjustable volume setting allows precise dispensing of samples with ease and confidence. OmniPette™ Single channel pipette for accurate and reliable pipetting results in your research projects.