CellPro™ Transfer or Electro Blotting buffer, 10X, pH 8.5, Sterile
Supplier: Alkali Scientific
DB0242
104026-706EA 59.98 USD
104026-706
CellPro™ Transfer or Electro Blotting buffer, 10X, pH 8.5, Sterile
Buffers Electrophoresis Buffers Transfer Buffers
Achieve efficient protein transfer with CellPro™ transfer or electro blotting buffer, offered in 10X concentration, pH 8.5, sterile and filtered.
  • Allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages
CellPro™ Transfer or electro blotting buffer, 10X, pH 8.5, provides a sterile and concentrated solution optimized for Western blotting and protein transfer applications. With a pH of 8.5, this buffer facilitates efficient protein transfer from polyacrylamide gels to membranes, ensuring accurate and reproducible results. The 10X concentration allows convenient dilution to achieve the desired working concentration, saving time and effort in the laboratory. Sterilized to maintain purity and eliminate the risk of contamination, CellPro™ transfer or electro blotting buffer is ready for immediate use in your protein research experiments.