Radiant™ Probe 1-Step Lo-Roxx qPCR Virus Kit
Supplier: Alkali Scientific
QL4099
104026-642EA 372.29 USD
104026-642
Radiant™ Probe 1-Step Lo-Roxx qPCR Virus Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Detect viral targets efficiently with Radiant™ Probe 1-Step Lo-Roxx qPCR Virus kit, designed for probe-based quantitative PCR.
  • Recommended kit for SARS-CoV-2 diagnostics
  • Validated for qualitative detection of SARS-CoV-2 nucleic acid
  • Concentrated 4X mix format, ideal for high-throughput, highly multiplexed assays
  • Ultra-sensitive detection of RNA viruses
  • Earlier detection of a wide range of template amounts
  • Includes UltraScript RTase for thermostable reverse transcription up to 55 °C
  • Advanced RNase inhibitor
  • Antibody-mediated hot start technology
  • Compatible with all real-time PCR platforms - standard and fast cycling conditions
cDNA Synthesis and qPCR in a single tube. Highly concentrated 4X mix allows increased sample and viral input. This optimizes sample sensitivity with small reaction volumes. Engineered for use with a wide range of probe technologies including TaqMan®, Scorpions® and molecular beacon probes.
Mix includes the thermostable Radiant™ Reverse transcriptase which is blended with an advanced RNase inhibitor to prevent degradation of RNA by contaminating RNase.