Powerblanket® Bucket Heaters
Supplier: POWERBLANKET LLC MS
BH05PRO BH05RR
104018-932EA 616 USD
104018-932 104018-952
Powerblanket® Bucket Heaters
Heaters Heating Mantles
Powerblanket’s 5-gallon bucket heaters provide a range of heating solutions for warming and storing temperature-sensitive materials. Our patented design eliminates hot and cold spots by applying uniform full-coverage heat across the entire surface of the container, allowing users to keep their materials at the proper viscosity while remaining at specified thermal levels. Each drum heater can be equipped with either an adjustable or a fixed thermostat.
  • Deliver uniform heat to temperature-sensitive products
  • Preserve expensive materials without scorching or burning
  • Prevent product waste by maintaining consistent temperatures
  • Cinch straps to secure tight fit
  • Safe for outdoor use
  • Water-resistant
  • Hassle free, preset internal 100 °F temperature on RR Rapid Ramp models
  • Hassle free, preset internal 145 °F temperature on PBL05 insulated band heater model
5-gallon bucket heaters come in two voltages: 120 volts and 240 volts. All of the 5-gallon bucket heaters come as an insulated full-wrap design with cinch straps to secure a tight fit. They’re also water-resistant and can be used on poly and steel buckets.
All of our 5-gallon bucket heaters come as an insulated full-wrap design with cinch straps to secure a tight fit. They’re also water-resistant and can be used on poly and steel buckets.
Powerblanket Lite - designed for light residential and industrial applications, low power density. Powerblanket - designed for any definitive heating applications, surrounded by a rugged vinyl shell, safe to use in temperatures as low as −20 °F. Powerblanket Xtreme - designed for harsh climates, safe to use in temperatures as low as −40 °F.
Adjustable temperature from ambient to 145 °F on PRO models. It is safe to use on both steel and poly buckets and provide an insulated full-wrap design making it more efficient.
Certifications: All models are certified by ETL to UL, CSA, and CE safety standards.