Powerblanket’s 275-gallon Hazardous Area Drum & Tote Heaters allow for simple, proper storage and heating for a variety of industrial supplies and chemicals. market.
- Hassle free, preset internal 80 °F temperature (±10 °F) / 27º C (±5 °C)
- Water resistant and safe for both steel & poly drums
- Insulated full-wrap design and cinch straps to secure tight fit
- Safe for outdoor use
- 33-foot cord (Plug not included)
Complete with special wiring and safety equipment to ensure the highest standards of safety, 275-gallon Hazardous Area Drum & Tote Heaters are Certified for C1D2 hazardous locations and explosive materials. Protect your critical materials, improve viscosity, and rest easy as Powerblanket solves your heating problems with complete safety.
Electrical heating equipment can cause explosions when used in areas with explosive substances, requiring other heating solutions. While band heaters provide good protection, the heat source is localized and can lead to uneven heating, hot spots, cold spots, and poor flow characteristics for certain materials. To prevent this, Powerblanket has developed the most advanced hazardous area heating systems on the market.
Certifications: Certified for C1D2 hazardous locations and explosive materials.