Triolein ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glyceryl trioleate, Glycerine trioleate, Glycerol Trioleate

331810100MG
100278-300EA 255 USD
100278-300 104018-212
Triolein ≥99%
Triolein

Store in a cool, dry place

Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
MDL Number: MFCD00137563
CAS Number: 122-32-7
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR