C58C1 Electrocompetent Agrobacterium, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1286-06 1286-18
104013-314PK 229.62 USD
104013-314 104013-316
C58C1 Electrocompetent Agrobacterium, Intact Genomics
Competent Cells
Intact Genomics C58C1 Electrocompetent Agrobacterium cells are useful for transgenic operations that involve potatoes, astragalus, rosaceae, tobacco and other plants.
  • High transformation efficiencies
  • Especially useful for cDNA gDNA library construction
Intact Genomics C58C1 electrocompetent Agrobacterium cells are optimized for the highest transformation efficiency and are useful for various applications. The chromosomal background of C58C1 is C58. C58 is cured of the Ti plasmid pTiC58 resulting in C58C1. C58C1 Competent cells may be useful for transgenic operations that involve Arabidopsis and other plants. This Agrobacterium strain is streptomycin and rifampicin resistant.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.