Anti-NS5A Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4856R-CY3
10399-138EA 823.27 USD
10399-138
Anti-NS5A Rabbit Polyclonal Antibody (Cy3®)
Antibodies
HCV is classified into the genus Hepacivirus of the family Flaviviridae. Like all the members of the family, HCV is an enveloped, single-stranded, positive-sense RNA virus. Its genome (about 9600 nt) is flanked at both termini by conserved, highly structured non-translated regions (NTRs) and encodes a polyprotein precursor (about 3000 aa), which is proteolytically processed by host and viral proteases to produce the structural (core, E1, E2 and p7) and non-structural (NS2, NS3, NS4A, NS4B, NS5A and NS5B) proteins of the virus. Recently, an additional protein has been identified, whose function remains unknown. NS5A is a ~56 kDa pleiotropic protein with key roles in both viral RNA replication and modulation of the physiology of the host cell. It's exact role is not currently known (2008).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NS5A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR