Streptococcus pyogenes Recombinant M1 CRISPR-Cas9 Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40572-A08B-200
103918-570EA 351.48 USD
103918-570
Streptococcus pyogenes Recombinant M1 CRISPR-Cas9 Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the s. pyogenes serotype M1 cas9 (WP_010922251.1) (Met1-Asp1368) was expressed with a nuclear localization sequence (NLS) at the N-terminus, two NLSs and a polyhistidine tag at the C-terminus.