SARS-CoV-2 (2019-nCoV) Spike S2 ECD-Fc Recombinant Protein (from HEK293 Cells)
Supplier: Sino Biological
40590-V02H-100
103886-030EA 452.48 USD
103886-030
SARS-CoV-2 (2019-nCoV) Spike S2 ECD-Fc Recombinant Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S2 ECD) (YP_009724390.1) (Ser686-Pro1213) was expressed with the Fc region of human IgG1 at the C-terminus.