Pair-N-Share Tandem Co-Culture Wells, Bulldog Bio
Supplier: Bulldog Bio
ARPS001
103881-420EA 1315.38 USD
103881-420
Pair-N-Share Tandem Co-Culture Wells, Bulldog Bio
Cell Culture Media
The Pair-N-Share offers a new way to look at cell-to-cell communication using a novel co-culture approach.
A 'trans-well' design that lets you 'see' the advantages
The Pair-N-Share is no ordinary 'bucket-in-a-bucket' trans-well co-culture vessel. Because its wells sit side by side, you can observe changes in both wells (on either side of the filter) with a standard microscope. The old method 'stacked' cells on two different substrates, making growth, communication, and observation less consistent and not easily observable.
The Pair-N-Share guarantees that cells can grow on identical surfaces in identical conditions, so no guesswork is needed as to the effects of matrix and positioning. The Pair-N-Share cultures grow and communicate side by side, with an optional filter restricting the flow of unwanted material. You can see the effects of intercellular communication in real time!