Tek-Select® Bouin’s Fixative, IMEB
Supplier: IMEB INC MS
S129
103783-074EA 232.31 USD
103783-074
Tek-Select® Bouin’s Fixative, IMEB
Stains Microscopy and staining reagents
Tek-Select® Bouin’s Fixative is ideally suited for gastrointestinal tract biopsies because it allows crisper and better nuclear staining than 10% neutral-buffered formalin.
  • Crisper and better nuclear staining
  • Packaged in one gallon bottle
This fixative is also excellent when tissue structure with a soft and delicate texture must be preserved.