Tek-Select® Diff-Stain Kit, IMEB
Supplier: IMEB INC MS
K7128
103783-058CS 283.08 USD
103783-058
Tek-Select® Diff-Stain Kit, IMEB
Stains Hematology Stains
The Tek-Select® Diff-Stain kit is designed to provide Nuclear and cytoplasmic staining and recently has provided excellent staining for Helicobacter pylori as well as a Hematology stain.
  • 8 oz. bottle of Solution A (fixative)
  • 8 oz. bottle of Solution B (azure stain)
  • 8 oz. bottle of Solution C (xanthene dyes)
  • Three 2 oz. staining containers
  • Storage tray
The Diff-Stain kit results in Nuclear staining from blue to purple, and pink to orange with paraffin and frozen section staining. Helicobacter pylori provides a deep rich purple. This stain is also excellent as a hematology stain.
Ordering information: Kit includes 8 oz. bottles of solution A (fixative), solution B (azure Stain), solution C (xanthene dyes), three 2 oz. staining containers, and a storage tray.