Bsu DNA Polymerase Large Fragment (from Bacillus subtilis)
Supplier: INTACT GENOMICS, INC MS
3585
103753-200EA 259.62 USD
103753-200
Bsu DNA Polymerase Large Fragment (from Bacillus subtilis)
Enzymes
Offers the largest collection of T4 Enzymes and proteins in the industry.
  • Extremely high purity
  • Variety of uses including RPA optimization
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 10X Bsu DNA polymerase I reaction buffer
Bsu DNA polymerase I, large fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5' - 3' polymerase activity of DNA polymerase I but lacks the 5' - 3' exonuclease activity. This large fragment also lacks 3' - 5' exonuclease activity.