T4 DNA Ligase, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
3212
103753-180EA 225 USD
103753-180
T4 DNA Ligase, Intact Genomics
Nucleic Acid Reagents Cloning Kits and Ligation
T4 DNA ligase displays significantly higher ligation efficiency than the nearest competitor.
  • 3 to 5X higher ligation efficiency than the nearest competitor
  • Extremely high purity
  • Useful for next-gen library preparation, cloning of PCR products, self-circularization of linear DNA and more.
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product.
  • The physical purity of this enzyme is ≥99% as assessed by SDS-PAGE with Coomassie® blue staining.
  • 10X T4 DNA ligase reaction buffer
T4 DNA ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. This enzymes joins DNA fragments with either cohesive or blunt termini as well as repair single stranded nicks in duplex DNA, RNA or DNA/RNA hybrids.
Delivery information: All our products are shipped on dry ice.