igTaq DNA Polymerase 2X Master Mix, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
3249
103753-148EA 161.54 USD
103753-148
igTaq DNA Polymerase 2X Master Mix, Intact Genomics
Nucleic Acid Reagents End-point PCR Enzymes and Kits
High quality Taq DNA polymerase 2X master mix useful for routine PCR applications.
  • Useful for routine PCR, primer extension, colony PCR and more
  • Formulated for amplifying long target DNA
  • Supplied as convenient master mix
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Taq DNA polymerase 2X premix with dye
  • 5X magic enhancer
Intact Genomics Taq DNA Polymerase 2X premix with dye is ready to use, containing Taq DNA polymerase, dNTPs, MgCl₂, and stabilizers. Taq DNA polymerase is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity. This product is supplied with 10X PCR reaction buffer, containing MgCl₂, which produces a final Mg²⁺ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3' ends.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.