Hot Start Taq 2X Master Mix, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
3296
103753-142EA 242.31 USD
103753-142
Hot Start Taq 2X Master Mix, Intact Genomics
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot start Taq 2X master mix reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.
  • Useful for routine PCR, primer extension, colony PCR and more
  • Supplied as convenient master mix
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • 5X magic enhancer
Hot start Taq DNA polymerase 2X master mix is ready to use premix which contains hot start Taq DNA Polymerase, dNTPs, MgCl₂ and stabilizers with optimized reaction buffer. Hot start Taq is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity. Hot start Taq DNA polymerase is chemically modified that leads to complete inactivation of the polymerase until the initial heat activation step at the start of PCR. Hot start PCR reduces non-specific amplification during setup stages of the reaction and helps increase PCR specificity and sensitivity.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.