DL39 (DE3) Chemically Competent Cells, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1061-12 1061-24
103753-098EA 298.85 USD
103753-098 103753-100
DL39 (DE3) Chemically Competent Cells, Intact Genomics
Competent Cells
Chemically competent DL39 (DE3) E. coli cells are specific for transformation and protein expression in order to uniformly and specifically label e.g. phenylalanine or leucine residues. DL39 (DE3) can also be used to reduce NMR cross-labeling via. transaminase activity for valine, leucine, isoleucine, aspartate, phenylalanine, tyrosine and tryptophan residues.
  • DL39 (DE3) chemically competent cells provide the highest transformation efficiency in the industry
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Includes DL39 (DE3) competent cells, pUC19 control DNA and recovery medium
Genomic features deficient in the aromatic (TyrB), branched-chain (JIvE), and aspartate (AspC) transaminases. Modified to contain the T7 expression system.
Delivery information: All our products are shipped with dry ice.