Endotoxin Removal System, EndoTrap® HD, Biovendor
Supplier: BioVendor
LET0010
103737-146EA 1398.29 USD
103737-146
Endotoxin Removal System, EndoTrap® HD, Biovendor
Buffers Electrophoresis Buffers Washing Buffers
EndoTrap® HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples.
  • Excellent recovery rate
  • Designed to meet the needs of pharma and biotech industry
  • Efficient removal of endotoxins in biomanufacturing processes
EndoTrap® HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate.
The EndoTrap HD has an endotoxin binding capacity of >5000000 EU/ml. EndoTrap removes more than 99.99% of LPS and has a sample recovery rate of >95%. The non-toxic bacteriophage-derived ligand so that there are no cytotoxic effects.
EndoTrap® HD is based on hydrophilic, dimensionally stable affinity matrix with excellent pressure/flow characteristics. A EndoTrap® Leakage ELISA is available for the quantitative determination of the EndoTrap® ligand leakage.
Ordering information: EndoTrap® HD 5/1 Endotoxin Removal System includes 5x1mL columns, 125mL 5x Equilibration Buffer, 125mL 5x Regeneration Buffer, and 125mL 5x Storage Buffer. Developed by Hyglos and manufactured by LIONEX.