Lithium chloride 2M in ethanol

Supplier: AQUA SOLUTIONS MS
Danger

FIN028A
103724-042EA 90.98 USD
103724-042
Lithium chloride 2M in ethanol
Lithium chloride
Formula: LiCl
MDL Number: MFCD00011078
CAS Number: 7447-41-8
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR