Human Recombinant CD22 (from HEK293 cells), His Tag
Supplier: Sino Biological
11958-H08H-100 11958-H08H-1
103612-904EA 198 USD
103612-904 103624-250 76642-144
Human Recombinant CD22 (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human CD22 isoform-beta (P20273-1) extracellular domain (Met 1-Arg 687) was fused with a polyhistidine tag at the C-terminus.