Human Recombinant PCSK9/NARC1 (D374Y, V474I, G670E) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10594-H08H1-50 10594-H08H1-500
103621-336EA 116.1 USD
103621-336 103621-338 76640-938
Human Recombinant PCSK9/NARC1 (D374Y, V474I, G670E) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human PCSK9 (NP_777596.2, with mutation Asp 374 Tyr, natural mutation Val 474 Ile and Gly 670 Glu) (Met 1-Gln 692) was expressed with a polyhistidine tag at the C-terminus.