Anti-RNASE1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
13468-T08-100
103588-010EA 142.96 USD
103588-010 103610-490 103610-492
Anti-RNASE1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human RNase A / Ribonuclease A / RNASE1 (rh RNase A / Ribonuclease A / RNASE1; Catalog#13468-H08H; P07998; Met1-Thr156). RNase A / Ribonuclease A / RNASE1 specific IgG was purified by Human RNase A / Ribonuclease A / RNASE1 affinity chromatography.

Type: Primary
Antigen: RNASE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR