Anti-SRPK1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2923R-CY5.5
10355-042EA 823.27 USD
10355-042
Anti-SRPK1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Plays a central role in the regulatory network for splicing, controlling the intranuclear distribution of splicing factors in interphase cells and the reorganization of nuclear speckles during mitosis. Hyperphosphorylates RS domain-containing proteins such as SFRS1, SFRS2 and ZRSR2 on serine residues during metaphase but at lower levels during interphase. Locks onto SFRS1 to form a stable complex and processively phosphorylates the RS domain. Appears to mediate HBV core protein phosphorylation which is a prerequisite for pregenomic RNA encapsidation into viral capsids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRPK1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR