Purge Gas Generator, PG14 L, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
45-1010
103549-820EA 5443.09 USD
103549-820
Purge Gas Generator, PG14 L, Peak Scientific
Gas Generators Carbon Dioxide Generators
The PG14L gas generator from Peak Scientific is suitable for laboratory detectors requiring CO2 free gas such as FT-IR analysis.
  • Purge Gas (PG) generators from Peak Scientific produce air which is free of CO₂
  • Ideal for commercial laboratories with a high throughput rate of samples
  • High performance filters and PSA ensure maximum reduction of moisture (< -70 °C dew point).
  • Few moving parts means minimum maintenance requirements and 24/7 performance
  • Position generator where required, wall mountable for small laboratories
  • More cost effective than any other gas supply as the FT-IR detector needs a constant flow of gas
  • Completely silent in operation
  • Can be supported by Precision Air Compressor
  • 12 month full comprehensive on-site warranty
The PG14L gas generator from Peak Scientific is suitable for laboratory detectors requiring CO₂ free gas such as FT-IR analysis. Using Pressure Swing Adsorption technology to remove organic contaminants and moisture from air, these generators provide a consistent flow of air to ensure little background noise in laboratory analysis.
Requiring minimal maintenance, the PG14L does require an external source of air to operate and can be supported by the Precision Air Compressor.
Gas oulet fittings: ¼" BSP female
Min/Max inlet pressure: 100/120 psi (6.9/8.3 bar)