Nitrogen Generator, NG5000 ultra high purity, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
10-2330
103549-814EA 19483.97 USD
103549-814
Nitrogen Generator, NG5000 ultra high purity, Peak Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The NG5000 delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC, DSC and DMA applications as well as other laboratory applications requiring up to 5L/min of ultra high purity nitrogen.
  • Suitable for applications such as GC, DSC, DMA
  • Suitable for applications requiring ultra high purity
  • Available with or without an external compressor
  • Contains self-regenerating Carbon Molecular Sieve column
  • Gas is supplied on demand so generator works to your schedule
  • 12 month comprehensive on-site warranty
The NG5000 delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of up to 5 L/min for GC, DSC and DMA applications as well as other laboratory applications requiring up to 5L/min of ultra high purity nitrogen. This generator requires a high quality source of in-house air to supply the unit. For laboratories without this air supply the NG5000(A) contains an internal compressor.
To reach their ultra high purity nitrogen output these generators use Pressure Swing Adsorption and Carbon Molecular Sieve. These separation technologies remove oxygen and other impurities in air, delivering hassle-free, ultra high purity nitrogen, on-demand for the laboratory.